CELLSTAR® CELLreactor™, Polypropylene Filter Top Tubes, Greiner Bio-One
Supplier: Greiner Bio-One
227245
89217-614CS 1453.28 CAD
89217-614
CELLSTAR® CELLreactor™, Polypropylene Filter Top Tubes, Greiner Bio-One
Filters Bottle Top Filters
The new CELLSTAR®CELLreactor™ can be used as small bioreactor for suspension and spheroid cell culture, facilitating miniaturization of large scale setups and maximizing the number of parallel experiments.
  • PTFE coating on caps helps guarantee maximum sterility
  • Conical design to fit in standard 50 ml centrifuge rotors
  • Free of detectable DNase, RNase, human DNA, Non-pyrogenic, Non-cytotoxic
Each CELLreactor™ cap features 8 holes and a specific USP VI certificated, PTFE coated capillary pore filter membrane with a pore size of 0.2 μm to guarantee maximalMore Product Information