CMUV Large Volume Metal Heating Mantles, Electrothermal
Supplier: Antyila Scientific
36013-17 36013-19 36013-23
89217-416EA 6683.87 CAD
89217-416 89217-418 89217-424 89217-426 89217-432 89217-434
CMUV Large Volume Metal Heating Mantles, Electrothermal
Heaters Heating Mantles
CMUV Electromantles deliver the benefits of the CMU Electromantle, but do so for very large 60° funnels, as well as pear-shaped and round bottom flasks
  • Bottom opening accommodates 60° funnels of various diameters, and also accepts large capacity round bottom and pear-shaped flasks (10 - 22 L).
  • Complete heating control is provided by 3 separate electrical circuits that enable independent top, middle, and bottom heating, each controlled with its own dial switch.
  • Units ensure spill-proof protection against electric shocks via a grounded earth stainless
More Product Information