VWR® Test Tube Racks
Supplier: VWR International
89215-716 89215-718 89215-720 89215-722 89215-724 89215-726 89215-728 89215-730 89215-732 89215-734 89215-736 89215-738 89215-740 89215-742 89215-744 89215-746 89215-748 89215-750 89215-752 89215-754 89215-756 89215-758 89215-760 89215-762 89215-764 89215-766 89215-768 89215-770 89215-772 89215-774 89215-776 89215-778 89215-780 89215-782 89215-784 89215-786 89215-788 89215-790 89215-792 89215-794 89215-796 89215-798 89215-800 89215-802 89215-804 89215-806 89215-808 89215-810 89215-812 89215-814 89215-816 89215-818 89215-820 89215-822 89215-824 89215-826 89215-828
89215-716CS 264.38 CAD
89215-716 89215-718 89215-720 89215-722 89215-724 89215-726 89215-728 89215-730 89215-732 89215-734 89215-736 89215-738 89215-740 89215-742 89215-744 89215-746 89215-748 89215-750 89215-752 89215-754 89215-756 89215-758 89215-760 89215-762 89215-764 89215-766 89215-768 89215-770 89215-772 89215-774 89215-776 89215-778 89215-780 89215-782 89215-784 89215-786 89215-788 89215-790 89215-792 89215-794 89215-796 89215-798 89215-800 89215-802 89215-804 89215-806 89215-808 89215-810 89215-812 89215-814 89215-816 89215-818 89215-820 89215-822 89215-824 89215-826 89215-828
VWR® Test Tube Racks
Racks Tube Racks
VWR® Test Tube Racks are available in both nylon glass filled or polypropylene options.
  • Both glass filled or polypropylene options are easy to handle
  • Racks may be used in an incubator, fume hood, refrigerator, or for benchtop applications
  • Autoclavable
  • No assembly required
Racks are a single piece molded from either nylon 6 glass fiber reinforced material, or polypropylene.
Order Now
SPECIFICATIONS
Environmentally Preferable