Rectangular Low Profile Plastic Cup Sinks, Scientific Plastics
Supplier: Scientific Plastics
W39081-150 F164L081-150 W39081-154 F164L081-154 W39081-158 F164L081-158
89209-720EA 402.57 CAD
89209-720 89209-722 89209-724 89209-726 89209-728 89209-730
Rectangular Low Profile Plastic Cup Sinks, Scientific Plastics
Faucets and Sinks Sinks
Rectangular Cup sink Ideal for retro-fit applications and ADA low countertop fumehoods.
  • Beveled sides help reduce splash
  • Tapered bottom eliminates water pooling
Regular Cup Skin for mounting at the rear of the fume hood and Extended tailpiece allows for mounting near the front of the fume hood.
Order Now
SPECIFICATIONS
Environmentally Preferable