Rectangular Tight-Head UN Rated Poly Pail, New Pig
Supplier: New Pig
DRM892
89208-786EA 99.51 CAD
89208-786
Rectangular Tight-Head UN Rated Poly Pail, New Pig
Buckets
Durable, blow-molded HDPE construction has no seams to hold residue
  • Rectangular design stores easily and is more space efficient than conventional round designs
  • Design stores and stacks easily, more space efficient than conventional round designs
  • Opaque white color allows liquid-level reading while still blocking most light
  • Widely compatible for storing variety of liquids
  • UN3H1/Y1.8/100
Opaque white color allows you to instantly read liquid level while still blocking most light. Widely compatible for storing a variety of liquids. Stackable to maximize storage space. UN3H1/Y1.8/100.