EXP®2 Stem Trap Kits and Replacement Stems, 2.7 µm HALO®
Supplier: Optimize
15-03997-HA 15-03997-HB 15-03997-HD 15-03997-HE 15-03997-HF 15-04003-HA 15-04003-HB 15-04003-HD 15-04003-HE 15-04003-HF 15-04009-HA 15-04009-HB 15-04009-HD 15-04009-HE 15-04009-HF 15-04015-HA 15-04015-HB 15-04015-HD 15-04015-HE 15-04015-HF 15-04021-HA 15-04021-HB 15-04021-HD 15-04021-HE 15-04021-HF 15-03992-HA 15-03992-HB 15-03992-HD 15-03992-HE 15-03992-HF 15-04008-HA 15-04008-HB 15-04008-HD 15-04008-HE 15-04008-HF 15-04014-HA 15-04014-HB 15-04014-HD 15-04014-HE 15-04014-HF 15-04020-HA 15-04020-HB 15-04020-HD 15-04020-HE 15-04020-HF 15-03997-HN 15-04003-HN 15-04009-HN 15-04015-HN 15-04021-HN 15-03992-HN 15-04001-HA 15-04001-HB 15-04001-HD 15-04001-HE 15-04001-HF 15-04001-HN 15-04008-HN 15-04014-HN 15-04020-HN
97056-420EA 2566.17 CAD
97056-420 97056-422 97056-424 97056-426 97056-428 97056-430 97056-432 97056-434 97056-436 97056-438 97056-440 97056-442 97056-444 97056-446 97056-448 97056-450 97056-452 97056-454 97056-456 97056-458 97056-460 97056-462 97056-464 97056-466 97056-468 97056-474 97056-476 97056-478 97056-480 97056-482 97056-484 97056-486 97056-488 97056-490 97056-492 97056-494 97056-496 97056-498 97056-500 97056-502 97056-504 97056-506 97056-508 97056-510 97056-512 89208-084 89208-088 89208-102 89208-106 89208-110 89208-086 89208-090 89208-092 89208-094 89208-096 89208-098 89208-100 89208-104 89208-108 89208-112
EXP®2 Stem Trap Kits and Replacement Stems, 2.7 µm HALO®
Chromatography Columns
The entire EXP®2 stem trap and reusable holder are only slightly larger than a standard HPLC fitting. Its slim architecture allows it to easily fit into crowded instrument compartments or to connect directly to tightly-spaced injection ports.
  • Rated to 20000+ psi (1400+ bar)
  • Hand-tight and wrench-tight configurationwith auto-adjusting ZDV connection
  • Low-volume/low-dispersion cartridges
  • For nano flow applications
When tightened by hand, the EXP2 stem trap seals to 8700+ psi. All configurations incorporate wrench-flats to enable flawless sealing to 20000+More Product Information