OPTI-LYNX™ Guard Column Cartridges
Supplier: Optimize
11-03128-EA 11-03128-EB 11-03128-EC 11-03128-ED 11-03128-EE 11-03128-EF 11-03128-EG 11-03128-EH 11-03128-JD 11-03782-EA 11-03782-EB 11-03782-EC 11-03782-ED 11-03782-EE 11-03782-EF 11-03782-EG 11-03782-EH 11-03782-JD 11-02630-JD
89207-726PK 1536.93 CAD
89207-726 89207-728 89207-730 89207-732 89207-734 89207-736 89207-738 89207-740 89207-742 89207-744 89207-746 89207-748 89207-750 89207-752 89207-754 89207-756 89207-758 89207-760 89207-762
OPTI-LYNX™ Guard Column Cartridges
Chromatography Columns
The OPTI-LYNX™ guard column is a high performance, cartridge-based guard system, suitable for use with any analytical column.
  • Can be used with any brand of column
  • Cartridge based system
  • Low dispersion cartridge interface
  • ZDV holder connection
  • Less than 2 µl swept volume
  • Extends column life
  • Provides effective protection with negligible impact
  • Rated to 6,000 psi
Some cartridge-based guard column products offer convenience with a price: higher internal volumes. At Optimize Technologies, we refuse to compromise performance for anything. OPTI-LYNX™More Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable