OPTI-LYNX™ Guard Column Single Cartridges
Supplier: Optimize
11-02255-EA 11-02255-EB 11-02255-EC 11-02255-ED 11-02255-EE 11-02255-EF 11-02255-EG 11-02255-EH 11-02308-EA 11-02308-EB 11-02308-EC 11-02308-ED 11-02308-EE 11-02308-EF 11-02308-EG 11-02308-EH 11-02308-JD 11-02640-EA 11-02640-EB 11-02640-EC 11-02640-ED 11-02640-EE 11-02640-EF 11-02640-EG 11-02640-EH 11-02640-JD 11-02255-JD
77100-322EA 377.43 CAD
77100-322 77100-324 77100-326 77100-328 77100-330 77100-332 77100-334 77100-336 89207-688 89207-690 89207-692 89207-694 89207-696 89207-698 89207-700 89207-702 89207-704 89207-706 89207-708 89207-710 89207-712 89207-714 89207-716 89207-718 89207-720 89207-722 89207-724
OPTI-LYNX™ Guard Column Single Cartridges
Chromatography Columns
The OPTI-LYNX™ Guard Column is a high performance, cartridge-based guard system, suitable for use with any analytical column.
  • Can be used with any brand of column
  • Cartridge based system
  • Low dispersion cartridge interface
  • ZDV holder connection
  • Less than 2 µL swept volume
  • Extends column life
  • Provides effective protection with negligible impact
  • Rated to 6,000 psi
Some cartridge-based guard column products offer convenience with a price: higher internal volumes. At Optimize Technologies, we refuse to compromise performance for anything. OPTI-LYNX™More Product Information