Orion™ Star™ A211 pH Benchtop Meter, Thermo Scientific

Supplier: Thermo Fisher Scientific
STARA2110 STARA2115 STARA2116 STARA2114 STARA2117 STARA2118 STARA2119
89206-302EA 1796.8 CAD
89206-302 89206-304 89260-360 76076-754 76076-756 76076-758 76076-760
Orion™ Star™ A211 pH Benchtop Meter, Thermo Scientific
Electrochemistry Meters pH and Multi-Parameter Meters
Reliably measure pH, mV, ORP, and temperature with the Orion™ Star™ A211 pH Benchtop Meter for advanced lab analysis and printer or computer interfacing.

  • Perform up to a five point pH calibration using automatic buffer recognition of NIST or DIN buffer groups
  • Calibration editing option allows individual points to be fixed without a full recalibration
  • Auto-Read mode holds a stable reading, Continuous mode shows changing readings with hold option, and Timed mode logs data at specific intervals
  • Eliminate the need for magnetic stir bars using the meter-
More Product Information
Order Now

SPECIFICATIONS

Environmentally Preferable
pH range –2.000 - 20.000
pH resolution 0.1, 0.01, 0.001
pH accuracy ±0.002 pH
mV range ±2000.0 mV
mV resolution 0.1 mV
mV accuracy ±0.2 mV or ±0.05% of reading whichever is greater
Temperature resolution 0.1 °C (0.1 °F)
Temperature accuracy ±0.1 °C
Display Graphic LCD with backlight
Battery type Four AA (sold separately)
Battery life 800 hours
Power supply Universal AC adapter (included) or 4 AA batteries (sold separately)
Operating temperature range 5…45 °C (41…113 °F)
Data logging capacity 2000 with time/date stamp
Memory Non-volatile memory preserves data log, calibration log, and meter settings
Languages English, Spanish, French, Italian, German, and Chinese (Portuguese and Korean available with software update)
IP rating IP-54
Dimensions 24L×18W×11H cm (9.5×7.1×4.3")
Outputs USB, RS232
Input BNC (pH or ORP electrode), pin-tip (reference electrode), 8 pin MiniDIN (ATC temperature probe), stirrer input (stirrer probe)
Alarm High/low limit alarm, calibration due alarm
No. of channels 1
Calibration pH with calibration editing option, relative mV (RmV), ORP and temperature
Calibration points 1 point relative mV (RmV) mode; 1 point ORP mode; 1 - 5 points pH mode
Weight 0.9 kg (2 lbs.)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR