VWR® Refrigerated Circulating Baths, 240 V
Supplier: VWR International
89202-964 89202-968 89202-972 89202-976 89202-980 89202-988 89202-992 89202-996 89203-004 89203-012 89203-022 89203-024
89202-964EA 9614.28 CAD
89202-964 89202-968 89202-972 89202-976 89202-980 89202-988 89202-992 89202-996 89203-004 89203-012 89203-022 89203-024
VWR® Refrigerated Circulating Baths, 240 V
Circulating Thermostats Closed Circulator Systems Cooling Circulators
VWR® Refrigerated Circulating Baths offer precise temperature control, at temperature ranges from -40 to 200 °C (depending on the controller).
  • Easy-to-view LCD display
  • Precise temperature control
Available in a wide range of sizes, with three controller options, these refrigerated circulating baths are ideal for a variety of applications.
The large LCD display on the MX temperature controller is easily viewed across the laboratory. The Advanced Digital temperature controller offers a touch-pad, while the Advanced Programmable Temperature ControllerMore Product Information