VWR® Heated Circulating Baths, 240V Models
Supplier: VWR International
89202-928 89202-944 89400-972 89400-976
89202-928EA 5731.8 CAD
89202-928 89202-944 89400-972 89400-976
VWR® Heated Circulating Baths, 240V Models
Circulating Thermostats Open Circulator Systems Baths with Integral Circulators
VWR® Heated Circulating Baths offer precise temperature control, at temperature ranges from ambient +10° to 200°C (depending on the controller).
  • Easy-to-read LCD display
  • Programmable for up to ten 100-step programs
  • Available in a wide range of sizes
Available in a wide range of sizes, with three controller options, these heated baths are ideal for many applications.
The large LCD display on the MX temperature controller is easily viewed across the lab. The Advanced Digital temperature controller offers a touch-pad, while the Advanced ProgrammableMore Product Information