Pre-Cleaned Bottles, HDPE, Wide Mouth, Environmental Express
Supplier: Environmental Express
BPC1183 BPC1200 BPC1170 BPC1191 BPC3200 BPC3170 BPC3183 BPC1199 BPC3199 BPC3174 BPC1184 BPC3601 BPC1601 BPC1180 BPC1168 BPC3191 BPC1189
23050-070CS 91.12 CAD
23050-070 23050-098 23050-108 23050-224 23050-240 23050-268 23050-282 23050-344 23050-347 23050-364 23050-366 23050-372 23050-370 89202-798 89202-814 89202-846 23050-343
Pre-Cleaned Bottles, HDPE, Wide Mouth, Environmental Express
Bottles Roller Bottles
All containers meet one of two levels of pre-cleaning. Level III indicates that containers have not received an EPA washing treatment. Level I indicates each case of containers have been custody-sealed and lot number-labeled for traceability.
Level I pre-cleaning also includes a Certificate of Analysis indicating that the containers received full EPA quality assurance treatment.
Natural high-density polyethylene bottles are supplied with polypropylene caps with F-217 PE liner.
Choose preserved bottles, pre-filled with solutions for user safety and convenience.