VWR® Talon® Double Buret Holder and Support Stand
Supplier: Troemner
80863382 17683-258 89202-626 89202-634 89202-636 89202-638
17683-054EA 419.15 CAD
17683-054 17683-258 89202-626 89202-634 89202-636 89202-638
VWR® Talon® Double Buret Holder and Support Stand
Retort Stands
Double buret clamp holds any size buret from micro to 100 mL capacity.
  • Electropolished stainless steel finish
  • Graduated markings
  • Easily adjusted height
  • Stainless steel or aluminum support rod
Simply compress the scissor-like mechanism, insert buret, and gently release to grip. Numbers and graduation on buret remain easy to read. For height adjustments, recompress mechanism and slide buret up or down and gently release. Clamp unit attaches to optional standard support rod with built-in reinforced hook connector. Adjusting nut faces forward for easy use.More Product Information