ECO Heating Circulators with Stainless Steel Bath, Lauda-Brinkmann
Supplier: Lauda-Brinkmann
L001191 L001192 L001194 L001195 L001196 L001197 L001198 L001200 L001201 L001202 L001171 L001172 L001174 L001175 L001176 L001177 L001178 L001180
89202-362EA 10094.38 CAD
89202-362 89202-364 89202-366 89202-368 89202-370 89202-372 89202-374 89202-376 89202-378 89202-380 89202-382 89202-384 89202-454 89202-456 89202-458 89202-460 89202-462 89202-464 89202-466 89202-468 89202-470 89202-472 89202-474 89202-476
ECO Heating Circulators with Stainless Steel Bath, Lauda-Brinkmann
Circulating Thermostats Closed Circulator Systems Heating Circulators
ECO current replacement for Ecoline
The intuitive menu navigation allows for easy operation. Both control heads are equipped with a mini-USB interface as a standard feature. Additional interfaces are available via the add-on modules. On-the-fly adjustment of the flow between internal and external circulation is made possible even during operation via the new flow rate switch. The more powerful units are equipped with the energy-saving LAUDA SmartCool system. All cooling thermostats are available with air-cooled or water-cooled options. Stainless steel bath.
There are innovations andMore Product Information