Odyssey® II Cleanroom Goggles, Sellstrom®
Supplier: Sellstrom
80230 S80231
89194-416EA 31.36 CAD
89194-416 89429-304
Odyssey® II Cleanroom Goggles, Sellstrom®
Goggles
Goggles with UV absorbing PC lens feature a flexible design to ensure a proper fit.
  • Goggles can be autoclaved
  • High temperature resistant
  • Complete panoramic vision
  • Ultra comfortable, soft goggle body for a snug seal
  • Non-vented body offers the best protection against liquids and dust
  • Synthetic Rubber Strap (Latex Free)
  • Easy strap adjustment
Store in a dry, cool place away from direct sunlight (<40°C).
They can be used comfortably over most prescription glasses.
Certifications: Meets ANSI Z87.1+ High Impact Standards, CUL certified to meet CAN/More Product Information