SevenGo Duo® pro™ SG98 pH/Ion/Optical Dissolved Oxygen Meter, METTLER TOLEDO®
Supplier: Mettler Toledo
51302662 51302663 51302664 51302661
89186-068EA 4831.9 CAD
89186-068 89186-070 89186-072 89186-066
SevenGo Duo® pro™ SG98 pH/Ion/Optical Dissolved Oxygen Meter, METTLER TOLEDO®
Electrochemistry Meters pH and Multi-Parameter Meters
Ideal for the determination for pH, mV, Ion concentration, and Dissolved Oxygen based on optical RDO® (Rugged Dissolved Oxygen) technology.
  • Multiparameter
  • Store up to 500 data points
  • Five-point calibration
  • Intelligent sensor management system (ISM®)
These meters are essential for special analyses, particularly in the in the environmental, chemical and food & beverage industry. The meter stores up to 500GLP measurements, with Date/Time, Sensor ID and Serial number, user and Sample ID. The meter features five point pH calibration with a choice ofMore Product Information