Durable Adhesive Vinyl Printer Labels, Brady
Supplier: Brady Worldwide
141990 141989 141988 141987 141994 141993 141992 141991 142002 142001 142000 141999 142014 142013 142012 142011 142010 142009 142008 142007 142018 142017 142016 142015 142026 142025 142024 142023 142031 142030 142029 142028 142027 142036 142035 142034 142033 142032 142195 142194 142197 142196 142199 142198 142185 142193 142184 142192 142183 142191 142182 142190 142046 142045 142038 142042 142189 142188 142047 142040 142043 142039 142041 142044 142037 142187 142186
89174-972RL 255.79 CAD
89174-972 89174-974 89174-976 89174-978 89174-980 89174-982 89174-984 89174-986 89174-988 89174-990 89174-992 89174-994 89174-996 89174-998 89175-000 89175-002 89175-004 89175-006 89175-008 89175-010 89175-012 89175-014 89175-016 89175-018 89175-020 89175-022 89175-024 89175-026 89175-028 89175-030 89175-032 89175-034 89175-036 89175-038 89175-040 89175-042 89175-044 89175-046 89175-048 89175-050 89175-052 89175-054 89175-056 89175-058 89175-060 89175-062 89175-064 89175-066 89175-068 89175-070 89175-072 89175-074 89175-076 89175-078 89175-080 89175-082 89175-084 89175-086 89175-088 89175-090 89175-092 89175-094 89175-096 89175-098 89175-100 89175-102 89175-104
Durable Adhesive Vinyl Printer Labels, Brady
Labels
B-595: Durable low-shrink vinyl with ultra-aggressive adhesive is ideal for labeling pipes, equipment, shelves and nearly any other industrial surface. This vinyl material with permanent acrylic adhesive keeps labels adhered in harsh climates and is resistant to chemicals, oils and detergents.
B-7569: Durable vinyl film is used for pipe markers, arrow tape, warehouse marking, floor marking, warning panels, safety signs and hazardous substance identification.