Cutters, Standard, Excelta
Supplier: Excelta
7210E+ 9240E 9241E 9243E 9244E 9245E 9246E 9248E 7243E 9240SL 9241E-AV 9241E-GM 9241S 9241SL 9241E-BO 9242SL 9243SL 9244E-GM 9244SL 9245SL 9246SL 9247S 9247SL 9248SL
63042-824EA 245.97 CAD
63042-824 89174-806 89174-808 89174-810 89174-812 89174-814 89174-816 89174-818 89174-820 89174-822 75880-700 76471-644 76471-646 76471-648 76471-650 76471-652 76471-654 76471-656 76471-658 76471-660 76471-662 76471-664 76471-666 76471-668 76471-670 76471-672
Cutters, Standard, Excelta
Cutters
These high-quality cutters are designed for cutting small soft wire up to 250,000 times without resharpening.
  • Three head shapes for various accessibility issues
  • Three types of cuts available
  • 'Set-Screw' stop prevents over-travel
  • Induction hardened blades for longevity
  • Ergonomic handles/dual leaf springs
These Lazer Line® side cutters are made of carbon tool steel with induction hardened cutting blades to RHC 63-65. The CNC machining allows the joint surfaces to be perfect to allow for long life. The dual leaf springs provide a smooth action. TheyMore Product Information