NORM-JECT® Luer-Lock and Luer-Slip Bulk Syringes, Two-Part, Nonsterile, Air-Tite
Supplier: Air-Tite
8300020458 8300020460 8300020453 8300027526 8300027527 8300027529 8300020456 8300020452
89213-178CS 1339.43 CAD
89213-178 89174-504 89174-506 89174-492 89174-494 89174-496 89174-498 89174-502 89174-491 89213-180 89213-182
NORM-JECT® Luer-Lock and Luer-Slip Bulk Syringes, Two-Part, Nonsterile, Air-Tite
Syringes General-Purpose Syringes
Norm-Ject® Syringes are latex free, contain no rubber, no silicone oil, styrene or DEHP and are DNA free.
  • All-plastic construction
  • Low background extractables
  • Wide solvent compatability
  • Positive safety stop to prevent accidental spills
The 5 ml syringes are graduated to 6 ml, 10 ml syringes are graduated to 12 ml, 20 ml syringes are graduated to 24 ml, and 50 ml syringes are graduated to 60 ml.
More biologically inert than standard 3-part syringes. Because of their composition they are indicated for chromatography and many laboratory procedures.