Bottle Rack, 20-Place BOD, Wheaton, DWK Life Sciences
Supplier: DWK Life Sciences (KIMBLE)
W227729
89174-478CS 228.17 CAD
89174-478
Bottle Rack, 20-Place BOD, Wheaton, DWK Life Sciences
Racks Bottle Racks
Manufactured from rugged and durable PVC coated wire, this rack is specifically designed to accommodate 60mL BOD bottles
Racks can be interlocked for safe transport or storage. Each rack offers a maximum capacity of 20 bottles.