CTC/LEAP Technologies GC Autosampler Syringes, Hamilton
Supplier: Hamilton
203084 203086 203189 203205 203363 203072 203197 203198 203043 203074 203076 203078 203080 203206 203209 203226 203219 203082 203185 203361 203225 203141 203181 203182 200740 200742 203362
82020-750EA 409.35 CAD
82020-750 82020-752 89173-280 89173-284 89173-286 89173-288 89173-290 89173-292 89173-294 89173-296 89173-298 89173-300 89173-302 89173-304 89173-306 89173-308 89173-310 89173-312 82020-748 89173-278 89368-702 89368-704 89368-706 89368-708 89368-710 10118-398 10118-400 10118-414
CTC/LEAP Technologies GC Autosampler Syringes, Hamilton
Syringes Syringes for Chromatography
The Hamilton C-Line syringe minimizes the potential for bent needles with a keyed flange design.
C-Line syringes incorporate several unique design features that ensure superior performance
An improperly installed syringe can cause mis-injections or bent needles due to poor alignment. The unique flange alignment design aids in installation and results in fewer bent needles while the plunger button can be adjusted to prevent plunger tip damage. An innovative direct attachment design of the needle to the barrel minimizes sample carryover. Hamilton's headspace syringe is specificallyMore Product Information