Corning® Reusable Plastic Volumetric Flasks, PP, Class B, Corning
Supplier: Corning
5641P-10 5641P-25 5641P-50 5641P-100 5641P-250 5641P-500 5641P-1L
89171-982CS 242.27 CAD
89171-982 89171-984 89171-986 89171-988 89171-990 89171-992 89171-994
Corning® Reusable Plastic Volumetric Flasks, PP, Class B, Corning
Flasks Volumetric Flasks
Made of PP, translucent, with tapered PP stopper.
  • With an individually calibrated graduation line
  • Recommended routine cleaning up to 60 °C will preserve graduations
  • Exposure to temperatures above 60 °C may affect accuracy
Reusable plastic flasks are manufactured to Class B tolerances as defined by ISO 1042.
The accuracy of volumetric ware depends on the care used in calibrating it, using the correct type of ware for the application, handling the ware properly and ensuring the ware is clean. Calibration of volumetric ware is usually done at 20 °CMore Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable