Transport Bags, 95 kPa, Therapak®
Supplier: Therapak, LLC
15650 15652 15654 15655 15674 15690
89170-946CS 440.07 CAD
89170-946 89170-948 89170-950 89170-952 89170-954 10497-174
Transport Bags, 95 kPa, Therapak®
Mailers and Shippers
95kPa transport bags are ideal to use as a secondary watertight receptacle in accordance with DOT and IATA regulations for shipping Biological Substances. DOT and IATA shipping regulations require that UN3373 Biological Substance, Category B specimens are transported in either a primary container or secondary packaging that has passed the 95kPa pressure differential test.
Each bag features a patented leak-resistant seal. Specimen tube(s) and other materials can be securely sealed within the front pocket of the bag by peeling away the white tape and folding the adhesive down to cover theMore Product Information