Absorbent Wadding for Cushioning Material, Therapak®
Supplier: Therapak, LLC
10501R
89170-940RL 475.03 CAD
89170-940
Absorbent Wadding for Cushioning Material, Therapak®
Mailers and Shippers
Protect fragile specimen shipments.
  • Wadding is 12 mm thick
  • 30 meter roll is perforated every 305 mm
This absorbent wadding is excellent filler and cushioning material for biological specimen transport.