Aqui-Pak™ Absorbent Pouches, Therapak®
Supplier: Therapak, LLC
10314 10316 10335G 10310
89170-934CS 984.47 CAD
89170-934 89170-936 10497-168 76203-692
Aqui-Pak™ Absorbent Pouches, Therapak®
Bags Sample Bags
Aqui-Pak™ was designed for the safe transport of laboratory specimen tubes by keeping the tubes separate and providing cushioning at the same time.
  • Ideal for Biological Substance Category B or Exempt Animal/Human Specimen packaging
  • Aqui-Pak™ pouches can be customized to individual specifications
It consists of a multi-layered polymeric material that has extremely high absorption properties. To use, simply place the tubes in the provided bays and insert the Aqui-Pak™ into secondary packaging. During shipment, Aqui-Pak™ is best used in combination withMore Product Information