Rtx®-CLPesticides Column Kit (0.32 mm Int.Ø), Restek
Supplier: Restek
11196
89169-004EA 2772.12 CAD
89169-004
Rtx®-CLPesticides Column Kit (0.32 mm Int.Ø), Restek
Chromatography Columns
Rtx®-CLPesticides columns are specially designed to overcome the coelutions and analyte breakdown typically encountered in chlorinated pesticide analyses for EPA methods 8081, 608, and CLP.
  • Application-specific columns for organochlorine pesticides and herbicides
  • Low bleed—ideal for high sensitivity GC-ECD or GC-MS analyses
  • Baseline separations in less than 10 minutes
  • Stable to 320 to 340 °C
  • Analyze EPA method 8081B, 8082A, 8151A, 504.1, 515, 508.1, and 552.2 compounds without time-consuming column changes
Improved resolution and faster analyses,More Product Information