VWR® Self-Seal Sterilization Pouches
Supplier: VWR International
89140-800 89140-802 89140-804
89140-800CS 1015.12 CAD
89140-800 89140-802 89140-804
VWR® Self-Seal Sterilization Pouches
Bags Sterilization Pouches
Sterilization pouches are made of surgical-grade paper and blue polypropylene/polyester film.
  • Color-coded indicators on each pouch
  • Sterilizable by steam or ETO
Easy-to-open lip closure, prefold self-adhesive strip, and flat seals ensure seal integrity.
Certifications: Indicator ink conforms to ISO 11140-1. Manufactured in a facility with ISO 9001:2008, ISO 13485, and HACCP certifications.