Neptune® S³ Low Retention Pipette Tips
Supplier: Biotix
63302157 63300508 63302158 63302167 63300544 63302163 63302164 63302165 63300534 63302160 63300520 63302161 63302178 63300567 63302177
89140-096CS 1039.03 CAD
89140-096 89140-098 89140-100 89140-106 89140-128 89140-130 89140-884 89140-888 89140-892 89140-898 89168-732 89168-734 89168-736 89168-758 89168-760 89425-648
Neptune® S³ Low Retention Pipette Tips
Pipette Tips
The exclusive S³ polymer delivers to within 0.1% of indicated volume, eliminating sample hold-up and ensuring a more precise application.
  • Autoclavable
  • Good sustainability
  • Superior stability, firm placement
  • Maximum space saving design
Eco-friendly empty pipette tips trays are manufactured from recycled polypropylene. When combined with ESP™ reload systems, these tip trays reduce plastic waste by up to 90%.
This proprietary technology is essential for quantitative assays where sample retention can hinder accurate results. Universal pipette tips areMore Product Information