Sulforhodamine 101-PEO₈-propionic acid succinimydyl ester amine-reactive fluorescent dye
Supplier: Biotium
Synonyms: Texas Red-PEO8 SE, Sulphorhodamine 101-PEO8 -propionic acid succinimydyl ester, SR101-PEO8 SE, Sulforhodamine 101-PEO8 -propionic acid succinimydyl ester
90099
89139-698EA 264.37 CAD
89139-698
Sulforhodamine 101-PEO₈-propionic acid succinimydyl ester amine-reactive fluorescent dye
Sulforhodamine 101-PEO₈-propionic acid succinimydyl ester
SR101-PEO8 SE (Sulforhodamine 101-PEO8-propionic acid succinimydyl ester; Texas Red®-PEO8 SE) has an approximately 32 Angstrom water-soluble PEG spacer that separates the dye from the amine-reactive SE group.
  • λEx/λ Em= 583/603nm
  • Dark purple solid soluble in DMF or DMSO
  • Store at -20°C and protect from light, especially in solution
Texas Red is a registered trademark of Thermo Fisher Scientific.