NBD-Ethylenediamine fluorescent dye
Supplier: Biotium
90047
89139-598EA 343.49 CAD
89139-598
NBD-Ethylenediamine fluorescent dye
NBD-Ethylenediamine
The fluorescence of NBD is highly dependent on the environment; it is virtually nonfluorescent in water but becomes strongly green fluorescent in hydrophobic media. NBD-ethylenediamine can be readily coupled to activated carboxy groups or sulfonyl chlorides.
  • λEx/λEm (MeOH) = 466/535 nm
  • Yellow solid soluble in DMSO or DMF
  • Store at 4°C and protect from light