6-Carboxyrhodamine 6G hydrochloride single isomer ≥95% (by HPLC) amine-reactive fluorescent dye
Supplier: Biotium
Synonyms: 6-CR 6G, Carboxyrhodamine 6G hydrochloride, 6-isomer
90011
89139-536EA 212.13 CAD
89139-536
6-Carboxyrhodamine 6G hydrochloride single isomer ≥95% (by HPLC) amine-reactive fluorescent dye
6-Carboxyrhodamine 6G hydrochloride
6-CR6G (full name: 6-Carboxyrhodamine 6G, hydrochloride, single isomer) is the free acid form of 6-carboxyrhodamine 6G single isomer.
  • λEx/λ Em (MeOH) = 520/546 nm
  • Orange red solid soluble in water (pH 7), DMF or DMSO
  • Store at 4°C and protect from light