5(6)-Carboxyrhodamine 110-X succinimudyl ester ≥90% (by TLC) amine-reactive fluorescent dye
Supplier: Biotium
90003
89139-520EA 538.43 CAD
89139-520
5(6)-Carboxyrhodamine 110-X succinimudyl ester ≥90% (by TLC) amine-reactive fluorescent dye
5(6)-Carboxyrhodamine 110-X succinimudyl ester
5-(and-6)-Carboxyrhodamine 110-X, succinimidyl ester (5(6)-CR 110-X, SE) readily couples to a primary or secondary amine. The dye is a superior alternative to 5(6)-SFX because of its exceptional phototostability and fluorescence insensitivity to pH (4-9).
  • λEx/λ Em (MeOH) = 502/524 nm
  • Orange red solid soluble in DMSO or DMF
  • Store at -20°C and protect from light
Order Now
SPECIFICATIONS
Environmentally Preferable