NBD-PE membrane stain, for histology

Supplier: Biotium

Synonyms: N-(7-Nitrobenz-2-oxa-1,3-diazol-4-yl)-1,2-dihexadecanoyl-snglycero-3-phosphoethanolamine, triethylammonium salt

60025
89139-326EA 629.49 CAD
89139-326
NBD-PE membrane stain, for histology
NBD-PE

NBD-PE (N-(7-Nitrobenz-2-oxa-1,3-diazol-4-yl)-1,2-dihexadecanoyl-snglycero-3-phosphoethanolamine, triethylammonium salt) in combination with Rhodamine-DHPE (#60026) or Texas Red®-DHPE (#60027) has been used to study membrane fusion via fluorescence resonance energy transfer (FRET) (1-3). NBD-PE has also been used in photobleaching recovery measurement.


  • λEx/λEm (MeOH) = 463/536 nm
  • Orange solid soluble in chloroform
  • Store at -20°C and protect from light

Formula: C₄₉H₉₀N₅O₁₁P
MW: 956.24 g/mol
Storage Temperature: Freezer
CAS Number: 178119-00-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR