BCECF AM ester ≥95% (by HPLC) fluorescent pH indicator

Supplier: Biotium

51011 51011-1 51012
89139-242EA 311.07 CAD
89139-242 89139-244 89139-246
BCECF AM ester ≥95% (by HPLC) fluorescent pH indicator
BCECF AM ester

BCECF, AM ester is membrane-permeant and thus can be loaded into cells via incubation. This product is a mixture of three molecular species, all of which are readily hydrolyzed by endogenous esterases into a single BCECF free acid form once they are in the cells.


  • Cell permeability: Membrane permeant
  • Colors: Green
  • Absorption/Emission 508 nm/530 nm (for BCECF acid)
  • White solid soluble in DMSO and DMF
  • Store at -20°C and protect from light, especially in solution

Formula: C₄₂H₄₀O₂₁
Storage Temperature: Freezer
MDL Number: MFCD00036969
CAS Number: 117464-70-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR