4',6'-Diamidino-2-phenylindole dihydrochloride (DAPI dihydrochloride) used in nuclear staining

Supplier: Biotium

Synonyms: 2-(4-Amidinophenyl)indole-6-carboxamidine Dihydrochloride, 2-(4-Carbamimidoylphenyl)-1H-indole-6-carboximidamide dihydrochloride, DAPI.2HCl, 2-(4-Carbamimidoylphenyl)indole-6-carboximidamide Dihydrochloride, DAPI 2HCl

40011
89139-054EA 269.01 CAD
89139-054
4',6'-Diamidino-2-phenylindole dihydrochloride (DAPI dihydrochloride) used in nuclear staining
4',6'-Diamidino-2-phenylindole dihydrochloride (DAPI dihydrochloride)

DAPI (4′,6-Diamidino-2-Phenylindole, dihydrochloride) is a popular blue DNA dye that is used as a nuclear counterstain in fluorescence microscopy, chromosome staining and flow cytometry. The dye binds to the minor groove of dsDNA with approximately 20-fold fluorescence enhancement, with higher affinity for A-T rich regions. Also see DAPI dilactate (catalog no. 40009), a more water soluble form of the dye.


  • Classic blue nuclear counterstain for fixed cells
  • Can be used at higher concentrations to stain live cells
  • λEx/λEm (with DNA) = 358/461 nm
  • Off-white solid soluble in H2O
  • Store at 4°C and protect from light, especially in solution


DAPI can be used to stain live bacteria (gram-positive and gram-negative) but in live yeast the staining is weak and not nuclear.

Formula: C₁₆H₁₅N₅·2HCl
MW: 350.25 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD00012681
CAS Number: 28718-90-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR