Anti-IgG Donkey Polyclonal Antibody (CF™ 594)

Supplier: Biotium

20115 20115-1
89138-618EA 411.57 CAD
89138-618 89138-620
Anti-IgG Donkey Polyclonal Antibody (CF™ 594)
Antibodies
Type: Secondary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: CF™ 594
Epitope: Heavy and Light chain
Host: Donkey
Isotype: IgG
Reactivity: Mouse
Order Now

SPECIFICATIONS

Antigen symbol IgG
Environmentally Preferable
Antigen name Immunoglobulin G
Antibody type Secondary
Clonality Polyclonal
Conjugation CF® 594
Reactivity Mouse
Host Donkey
Isotype IgG
Western blot Yes
ELISA Yes
ImmunoFluorescence Yes
Flow cytometry Yes
Epitope Heavy and Light chain
Cross adsorption Minimal Cross-Reactivity: Bv, Ch, Gt, GP, Hs, Hu, Rb, Sh, SHm

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR