Anti-IgG Goat Polyclonal Antibody (CF™ 405S)

Supplier: Biotium

20080 20080-1
89138-540EA 377.32 CAD
89138-540 71003-996
Anti-IgG Goat Polyclonal Antibody (CF™ 405S)
Antibodies
Type: Secondary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: CF™ 405S
Epitope: Heavy and Light chain
Host: Goat
Isotype: IgG
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR