X-β-D-cellobioside

Supplier: Biotium

Synonyms: 5-Bromo-4-chloro-3-indolyl-β-D-cellobioside

10026
89138-246EA 258.39 CAD
89138-246
X-β-D-cellobioside
X-β-D-cellobioside
Formula: C₂₀H₂₅BrClNO₁₁
MW: 570.78 g/mol
CAS Number: 177966-52-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR