4-Methylumbelliferyl phosphate disodium salt (4-MUP-Na2)

Supplier: Biotium

10010 10010-1
89138-212EA 118.25 CAD
89138-212 89138-214
4-Methylumbelliferyl phosphate disodium salt (4-MUP-Na2)
4-Methylumbelliferyl phosphate disodium salt (4-MUP-Na2)
Formula: C₁₀H₇Na₂O₆P
MDL Number: MFCD00037614
CAS Number: 22919-26-2
EINECS: 245-325-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR