uTIP™ Pipette Tips for Universal Pipettors, Biotix
Supplier: Biotix
63300052 63300048 63300049 63300050 63300054 63300187 63300188 63300055 63300056 63300057 63300059 63300061 63300058 63300060 63300081 63300051 63300079 63300053 63305451 63305452 63305453 63305454
89136-566CS 726.95 CAD
89136-566 89136-570 89136-574 89136-578 89408-548 89425-644 89425-646 10027-666 10027-668 10027-670 10027-672 10027-736 76175-282 76175-382 76175-388 76175-408 76175-410 76175-414 77575-924 77575-926 77575-928 77575-930
uTIP™ Pipette Tips for Universal Pipettors, Biotix
Pipette Tips
Biotix® uTIP Racked- Non-Filtered, Low Retention, Pre-Sterilized Universal pipette Tips are designed to improve ergonomics by lowering the necessary insertion and ejection force by 66 to 80% to make a secure seal. This is achieved through the patented Flexfit™ and Blade™ technologies. All Biotix® tips are manufactured with X-Resin™, a proprietary ultra-low retention resin which results in tighter CV values.
  • The FlexFit™ seal is able to be secured with just the weight of a pipettor
  • Blade™ Technology lowers the chance of hanging droplet formation by
More Product Information
New Product
Order Now
SPECIFICATIONS
Environmentally Preferable