EcoPolyBlend DrumSheds™, Justrite®
Supplier: Justrite
28675 28676 28679
89134-686EA 7340.79 CAD
89134-686 89134-688 89134-694
EcoPolyBlend DrumSheds™, Justrite®
Drum Storage
Eco-friendly, weather-resistant drum storage.
  • Spacious interior for funnels or pumps
  • Leak-proof pallet sump meets EPA and SPCC requirements
  • Forklift pockets front and back for easy transport
  • Ships fully assembled
Weather-resistant outdoor sheds offer high performance spill control. A robust design made of 55% recycled polyethylene features convenient rolltop doors to allow access from front or rear. Forklift pockets on the front and back enable easy transport and relocations.The EPA/SPCC compliant leakproof pallet sump features thick, durable deckMore Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable