Type I Safety Cans for various types of liquids, Justrite®
Supplier: Justrite
7110200 7110210 7110300 7110310 7110400 7110410 7120200 7120210 7120300 7120310 7120400 7120410 7125200 7125210 7125300 7125310 7125400 7125410 7150200 7150210 7150300 7150310 7150400 7150410
89134-546EA 122.56 CAD
89134-546 89134-548 89134-550 89134-552 89134-554 89134-556 89134-562 89134-564 89134-566 89134-568 89134-570 89134-572 89134-578 89134-580 89134-582 89134-584 89134-586 89134-588 89134-596 89134-598 89134-604 89134-606 89134-608 89134-610
Type I Safety Cans for various types of liquids, Justrite®
Safety Cans
Compliant and economical Justrite™ Type I safety cans reduce fire risks and make everyday use of flammables easier and safer
Blue- kerosene, Yellow - diesel, Green-oils.
Protection features include a 3-1/2" (88mm) long chemical resistant stainless steel flame arrester that stops flashback ignition, provides faster liquid flow, and exceptional durability. The self-venting, self-closing leakproof lid prevents ruptures from pressure build-up and controls vapors and spills. Warning label includes an extra large ID zone for user labeling of contents to avoid misuse, reduce the potential forMore Product Information