Pig Blue® Absorbent Mat Roll, New Pig
Supplier: New Pig
BLU104 BLU105 BLU102 BLU103
89132-772EA 321.12 CAD
89132-772 89132-774 89132-776 89132-778
Pig Blue® Absorbent Mat Roll, New Pig
Spill Control Materials
Consistent construction produces a mat that's free of light spots to absorb liquid evenly for better wicking and retention
Contains 75% recycled fibers; ideal in situations where green products are desired or required. Fire retardant for added safety; flame-tested in accordance with ASTM Standard E84-08. Absorbs oils, coolants, solvents and water; not recommended for acids, bases or other corrosive liquids. Rolls are ideal for covering large areas and soaking up leaks and overspray. Easy-tear perforations let you take only what you need, so you use less mat and save money.
HeavyweightMore Product Information