VWR® Photo Imager
Supplier: VWR International
89131-412 89131-416 89131-420 89131-424 89131-428 89131-432 89131-436
89131-412EA 7573.68 CAD
89131-412 89131-416 89131-420 89131-424 89131-428 89131-432 89131-436
VWR® Photo Imager
Photodocumentation Systems
Capture color gel images with the Photo Imager’s digital color camera and print brilliant color 4x6” images in seconds.
  • High resolution digital camera to capture brilliant gel images
  • Lightweight hood that fits over transilluminators with filter sizes up to 21x26 cm (81/4x101/4")
  • Choice of UV transilluminator
  • Ethidium bromide filter positioned inside the hood
  • Viewport window with UV blocking to allow visibility of gels
  • Brilliant 10.2x15.3 cm (4x6”) color prints, 300 x 300 resolution, can be generated in seconds
The Photo Imager allows users toMore Product Information