Axygen® 96-Well PCR Plate, Corning
Supplier: Corning
PCR-96-FLT-C PCR-96-LP-FLT-C PCR-96-LP-AB-C PCR-96-LP-FLT-W PCR-96-LP-AB-W PCR-96-LC480-W PCR-96-LC480-W-BC PCR-96-LC480-W-NF PCR-96-FS-C-S
89005-550CS 1685.86 CAD
89005-550 89005-552 89005-554 89006-880 89006-884 89130-568 89130-570 89130-572 89179-632
Axygen® 96-Well PCR Plate, Corning
Microplates PCR Plates
Axygen's plates are designed for PCR applications.
  • RNase-/DNase-free
  • Nonpyrogenic
  • Nonsterile
  • Manufactured with 99.9% pure virgin polypropylene
Ultra-thin and consistent wall thickness allows precise thermal transfer for optimum results. Can be sealed with 8/12 strip caps, Axymat or sealing film.
Order Now
SPECIFICATIONS
Environmentally Preferable Low Manufacturing Impact
Environmental certification I-RECs, ISO 14001 - Reynosa