PolarSafe™ White and Colored Cardboard Freezer Storage Boxes And Dividers, Argos Technologies
Supplier: Antyila Scientific
06755-74 06755-75 06755-78 06755-80 06755-81 06755-79 06755-82 06755-83 06755-84 06755-85 06755-86 06755-71 06755-76 06755-72 06755-73 06755-77 04395-41 04395-40 04395-34 04395-35 04395-36 04395-37 04395-38
89128-176EA 12.32 CAD
89128-176 89128-178 89128-184 89128-186 89128-188 89128-190 89128-192 89128-194 89128-196 89128-198 89128-200 89128-202 71001-640 71001-642 71001-644 89128-183 10030-118 10030-120 10436-314 10436-316 10436-318 10436-320 10436-322
PolarSafe™ White and Colored Cardboard Freezer Storage Boxes And Dividers, Argos Technologies
Boxes Cryoboxes
PolarSafe™ cryo/freezer boxes and dividers are made of sturdy white cardboard, with a moisture-repellent outer coating to protect from the effects of liquid nitrogen or condensation from refrigerated equipment
The 81 and 100 place boxes have a moisture repellent lining on the inside of the box as well as on the dividers. In addition, both boxes have been printed with a unique numerical grid to keep your samples organized.
Two specialty boxes for PCR are available. The first is 5 x 5 x 1 and will accommodate a 196 place cardboard divider. The 1" height prevents the tubes from droppingMore Product Information