Argos Technologies® PolarSafe™ Stainless Steel Upright Freezer Racks, Cole-Parmer
Supplier: Antyila Scientific
04399-34 04400-69 04399-36 04399-41 04398-24 04399-20 04398-32 04399-28 04398-18 04398-12 04398-13 04398-37 04398-14 04398-15 04398-17 04398-19 04398-20 04398-53 04398-21 04398-22 04398-30 04398-31 04401-28 04401-29 04401-34 04401-42 04398-56 04399-26 04398-71 04398-16 04398-26 04398-59 04398-55 04398-23 04398-64 04398-25 04399-23 04399-29 04399-33 04399-65 04399-66 04399-67 04399-69 04399-70 04399-71 04401-27 04401-33 04401-35 04398-27 04401-38 04401-39 04401-40 04401-41 04401-44 04401-64 04401-65 04401-66 04401-67 04401-68 04401-69 04401-70 04398-33 04404-24 04404-25 04404-26 04404-27 04404-28 04404-30 04404-31 04404-33 04404-39 04404-40 04404-41 04404-42 04404-43 04404-44 04404-77 04404-78 04404-79 04404-80 04404-81 04404-82 04404-84 04404-85 04398-38 04398-73 04398-70 04398-69 04398-41 04398-62 04398-52 04398-57 04398-68 04400-68
71001-406EA 493.42 CAD
71001-406 71001-408 71001-410 71001-412 71001-414 71001-424 71001-426 71001-428 71001-430 89128-102 89128-104 89128-108 89128-110 89128-112 89128-116 89128-118 89128-120 89128-122 89128-124 89128-126 89128-146 89128-148 89128-156 89128-158 89128-164 89128-166 89128-168 89168-878 89168-882 89128-106 89128-114 89128-170 89220-466 89220-468 89220-470 89220-472 89220-474 89220-476 89220-478 89220-480 89220-482 89220-484 89220-486 89220-488 89220-490 89220-492 89220-506 89220-510 89220-512 89220-514 89220-516 89220-518 89220-520 89220-522 89220-524 89220-526 89220-528 89220-530 89220-532 89220-534 89220-536 89220-538 89220-692 89508-516 89508-518 89508-520 89508-522 89508-524 89508-526 89508-528 89508-530 89508-532 89508-534 89508-536 89508-538 89508-540 89508-542 89508-544 89508-546 89508-548 89508-550 89508-552 89508-554 89508-556 89508-558 89508-560 89508-562 89508-564 89508-566 89508-568 89508-570 89508-572 89508-574 89508-576 89508-578 89508-580 89508-620 89508-622 89508-624 89508-626 89508-628 76320-760 76320-894 76320-896 76320-898 76320-900 76320-902
Argos Technologies® PolarSafe™ Stainless Steel Upright Freezer Racks, Cole-Parmer
Racks Cryogenic Box Racks
Argos Technologies® PolarSafe™ Metal Freezer Storage Racks are constructed of high-quality, corrosion resistant stainless steel
Argos Technologies® PolarSafe™ Racks have been configured to fill all major ultra low (-80°) freezers on the market; racks are designed for Upright and Chest ultra low freezers.
Handles at both ends accommodate freezers with right or left handed doors. Our extensive product range will accommodate all type of sample containers including 2", 3", and 5" boxes, microtiter and deepwell plates and 15 & 50 mL centrifuge tubes.
Ordering information: AdditionalMore Product Information