VWR® TraceClean® Boston Round Glass Jars, Narrow Mouth, Amber
Supplier: VWR International
89126-556 89126-558 89126-560 89126-564 89126-566 89126-654
89126-556CS 124.3 CAD
89126-556 89126-558 89126-560 89126-564 89126-566 89126-654
VWR® TraceClean® Boston Round Glass Jars, Narrow Mouth, Amber
Jars General Use Jars
These TraceClean® glass jars are used primarily for the collection of aqueous media for testing semi-volatiles, pesticides, PCBs, diesel range organics, TPH, metals, and phenolics.
  • Amber glass fabrication
  • Protects UV light-sensitive contents
  • Narrow-mouthed design
Secondary uses include containment and archiving
Quality Assured® with VWR® Logo Sample ID Label products, a lot certificate of analysis is generated with the analytical results obtained including the concentration reporting limits and units of measurement. This certificate of analysis isMore Product Information