Biotix Disposable Reagent Reservoirs, Biotix
Supplier: Biotix
63300132 63300131 63300130 63300135 63300134 63300133 63300117 63300118 63300119 63300120 63300121 63300122 63300124 63300125 63300126 63300127 63300128 63300129
89125-668CS 140.61 CAD
89125-668 89125-670 89125-672 89125-674 89125-676 89125-678 89511-184 89511-186 89511-188 89511-190 89511-192 89511-194 89511-196 89511-198 89511-200 89511-202 89511-204 89511-206
Biotix Disposable Reagent Reservoirs, Biotix
Reagent Reservoirs
Biotix disposable reagent reservoirs are designed for use in multi-channel pipetting applications and can accommodate up to 12-channel pipettors. Angled sides and a V-shaped bottom allow for maximum recovery of reagents. All reservoirs feature internal graduated volume lines and pour-spouts on the corners. Choose from opaque white polyvinyl chloride (PVC) or high clarity polystyrene (PS) in several packaging configurations.
Biotix disposable reagent reservoirs are designed for use in multi-channel pipetting applications and can accommodate up to 12-channel pipettors
Biotix ReagentMore Product Information