EasyClean® 360 Isolator Cleaning System, Berkshire
Supplier: Berkshire
EC360.ICT.1 EC360.EC.6 EC360ECST20 EC360.NW.40
89125-662CS 738.89 CAD
89125-662 89125-664 89125-666 89136-880
EasyClean® 360 Isolator Cleaning System, Berkshire
Mops
For efficient cleaning of compounding isolators, biological safety cabinets, laminar flow hoods, and other mini-environments
  • Triangular head 4×7" (10×18 cm) shape allows for improved access in tight corners
  • Mop head and handles can be sterilized by autoclave or gamma irradiation
  • Stretch fabric and elastic edge allow easy installation and secure fit on the mop head
  • Covers are nonshedding and low-linting per USP <797> and USP <800> requirements
Durable stainless steel mop head swivels 360° for easy maneuverability. Head can be locked for 180°More Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable